Skip to Content
Merck
Styrene ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%

Styrene

Styrene ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Synonym(s):
Styrene, Phenylethylene, Vinylbenzene
Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique