
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride

Synonym(s):
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, L-α-(2-Aminoethoxyvinyl)glycine hydrochloride, AVG-HCl
Empirical Formula (Hill Notation):
C6H12N2O3 · HCl
CAS Number:
Molecular Weight:
196.63
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Technique
Showing 1-2 of 2