
(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

Synonym(s):
(S,S)-Jacobsen’s catalyst
Linear Formula:
[[[(CH3)3C]2C6H2-2-(O-)CH=N]2C6H10]MnCl
CAS Number:
Molecular Weight:
635.20
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing