
Squalane

Synonym(s):
2,6,10,15,19,23-Hexamethyltetracosane, Cosbiol, Perhydrosqualene, Robane, Squalane
Linear Formula:
[(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)CH2CH2-]2
CAS Number:
Molecular Weight:
422.81
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing