
Spironolactone Related Compound C

Synonym(s):
17α-(2-Carboxyethyl)testosterone γ-lactone
Empirical Formula (Hill Notation):
C22H30O3
CAS Number:
Molecular Weight:
342.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing