
Sodium taurochenodeoxycholate

Synonym(s):
2-([3α,7α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid sodium salt, 3α,7α-Dihydroxy-5β-cholan-24-oic acid N-(2-sulfoethyl)amide sodium salt, Taurochenodeoxycholic acid sodium salt
Empirical Formula (Hill Notation):
C26H44NNaO6S
CAS Number:
Molecular Weight:
521.69
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing