Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium tartrate dibasic dihydrate
Synonym(s):
L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate
Empirical Formula (Hill Notation):
C
4
H
4
Na
2
O
6
· 2H
2
O
CAS Number:
6106-24-7
Molecular Weight:
230.08
EC Number:
212-773-3
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (2)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S4797
Sodium tartrate dibasic dihydrate,
BioXtra, ≥99.0%
Expand
View Pricing
71995
Sodium tartrate dibasic dihydrate,
purum p.a., ≥98.0% (NT)
Expand
View Pricing