Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium sulfate decahydrate
Synonym(s):
Glauber’s salt, Sodium sulfate decahydrate
Linear Formula:
Na
2
SO
4
· 10H
2
O
CAS Number:
7727-73-3
Molecular Weight:
322.20
EC Number:
231-820-9
Products
Product Category
salts (6)
Brand
SAFC (1)
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
403008
Sodium sulfate decahydrate,
ACS reagent, ≥99.0%
Expand
View Pricing
246980
Sodium sulfate decahydrate,
reagent grade, 97%
Expand
View Pricing
71970
Sodium sulfate decahydrate,
puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Expand
View Pricing
1.06648
Sodium sulfate decahydrate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.06642
Sodium sulfate decahydrate,
crystalline, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JPE, USP
Expand
View Pricing
71969
Sodium sulfate decahydrate,
BioUltra, ≥99.0% (calc. on dry substance, T)
Expand
View Pricing