Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium selenate
Linear Formula:
Na
2
SeO
4
CAS Number:
13410-01-0
Molecular Weight:
188.94
EC Number:
236-501-8
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S0882
Sodium selenate,
≥95% (elemental analysis)
Expand
View Pricing
71948
Sodium selenate,
purum p.a., ≥98.0% (T)
Expand
View Pricing
S8295
Sodium selenate,
BioXtra
Expand
View Pricing