
Sodium phosphonoformate tribasic hexahydrate

Synonym(s):
Foscarnet, Phosphonoformic acid trisodium salt hexahydrate
Linear Formula:
Na2O3PCO2Na · 6H2O
CAS Number:
Molecular Weight:
300.04
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing