
Sodium monofluorophosphate

Synonym(s):
Sodium fluorophosphate
Linear Formula:
Na2FPO3
CAS Number:
Molecular Weight:
143.95
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1