Skip to Content
Merck
Sodium iminodiacetate dibasic monohydrate ≥95.0% (T)

Sodium iminodiacetate dibasic monohydrate

Sodium iminodiacetate dibasic monohydrate ≥95.0% (T)
Synonym(s):
Iminodiacetic acid disodium salt monohydrate
Linear Formula:
HN(CH2COONa)2 · H2O
CAS Number:
Molecular Weight:
195.08
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing