Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium gluconate
Synonym(s):
Sodium gluconate, D-Gluconic acid sodium salt, 2,3,4,5,6-Pentahydroxycaproic acid sodium salt, D-Gluconate sodium salt, Sodium D-gluconate
Empirical Formula (Hill Notation):
C
6
H
11
NaO
7
CAS Number:
527-07-1
Molecular Weight:
218.14
EC Number:
208-407-7
Products
Product Category
carbohydrates (1)
monosaccharides (1)
salts (1)
Brand
SAFC (1)
Sigma-Aldrich (1)
Biological Source
Color
Feature
Formula Weight
Physical Form
Agency
Quality Segment
Application
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
8.17054
Sodium gluconate,
E 576
Expand
View Pricing
S2054
Sodium gluconate,
meets USP testing specifications
Expand
View Pricing