Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium citrate dibasic sesquihydrate
Synonym(s):
Citric acid disodium salt, Disodium hydrogen citrate sesquihydrate
Linear Formula:
HOC(COOH)(CH
2
COONa)
2
· 1.5H
2
O
CAS Number:
6132-05-4
Molecular Weight:
263.11
EC Number:
205-623-3
Products
Brand
Sigma-Aldrich (1)
Formula Weight
pH Value
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
71635
Sodium citrate dibasic sesquihydrate,
purum p.a., ≥99.0% (T)
Expand
View Pricing