Skip to Content
Merck
Sodium acetate Redi-Dri™, free-flowing, anhydrous, ReagentPlus®, ≥99.0%

Sodium acetate

Sodium acetate Redi-Dri™, free-flowing, anhydrous, ReagentPlus®, ≥99.0%
Synonym(s):
Acetic acid sodium salt, Sodium acetate, OmniPur® Sodium Acetate, Anhydrous
Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing