Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SmartPak
®
Purification Pack
Products
Product Category
polishers (7)
Brand
Milli-Q (7)
Compatibility
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
SPR00SIA1
SmartPak
®
Purification Pack,
For use with Direct-Q
®
3 Systems
Expand
View Pricing
SPR0LSIA1
SmartPak
®
Purification Pack,
Purifies pure and ultrapure water for Direct-Q
®
systems with low (<15 °C) feed water temperature
Expand
View Pricing
SPR08SIA1
SmartPak
®
Purification Pack,
For Direct-Q
®
8 Water Purification Systems
Expand
View Pricing
SPR00SIA2
SmartPak
®
Purification Pack,
For RiOs-DI
®
Systems
Expand
View Pricing
SPR00S001
SmartPak
®
Purification Pack,
For use with RiOs
™
3 Systems
Expand
View Pricing
SPR08S001
SmartPak
®
Purification Pack,
For use with RiOs
™
8 Water Purification System
Expand
View Pricing
SPR0LS001
SmartPak
®
Purification Pack,
For use with RiOs
™
3/5 Water Purification System
Expand
View Pricing