
(S)-Lactic acid

Synonym(s):
L-(+)-Lactic acid, (S)-2-Hydroxypropionic acid, Sarcolactic acid
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing