Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sigma 7-9
®
Synonym(s):
Trizma
®
base, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Linear Formula:
NH
2
C(CH
2
OH)
3
CAS Number:
77-86-1
Molecular Weight:
121.14
EC Number:
201-064-4
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Purity
Research Area
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
T1378
Sigma 7-9
®
,
≥99% (titration), crystalline, Sigma 7-9
®
Expand
View Pricing