Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Septa, white PTFE/silicone
Products
Product Category
septa (4)
Brand
Supelco (4)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27361
Septa, white PTFE/silicone,
septum diam. × thickness × PTFE thickness 20 mm × 0.125 in. × 0.005 in., pkg of 100 ea
Expand
View Pricing
27356
Septa, white PTFE/silicone,
diam. × thickness 11 mm × 0.075 in., for use with 4 mL vial, pkg of 100 ea
Expand
View Pricing
27374
Septa, white PTFE/silicone,
diam. × thickness × PTFE thickness 20 mm × 125 mil × 0.005 in., for use with 20mm crimp seal, 22 mm cap, pkg of 1000 ea
Expand
View Pricing
27369-U
Septa, white PTFE/silicone,
diam. × thickness 11 mm × 0.075 in., for use with screw top vial, 4 mL, with 13-425 thread, pkg of 1000 ea
Expand
View Pricing