Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SEM Sample Holder
Products
Brand
Sigma-Aldrich (16)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
933120
SEM Sample Holder,
Vertical for Thin Sample
Expand
View Pricing
933325
SEM Sample Holder,
Multi Holder for 6 Pin Stubs, Hitachi, M4
Expand
View Pricing
933309
SEM Sample Holder,
45° Pin Stub Holder, size 3.2 mm , (1/8”) pin
Expand
View Pricing
933279
SEM Sample Holder,
Variable Tilt Mount holder, For Pin
Expand
View Pricing
933333
SEM Sample Holder,
24 Place SEM Holder
Expand
View Pricing
933317
SEM Sample Holder,
Multi Pin Stub Adapter, Jeol
Expand
View Pricing
933376
SEM Sample Holder,
SEM Pin Stub Adapter, size 12.5 mm
Expand
View Pricing
933368
SEM Sample Holder,
SEM Pin Stub Adapter, size 15 mm
Expand
View Pricing
933112
SEM Sample Holder,
EMS Orbital Specimen Holder, Pin Mount
Expand
View Pricing
933287
SEM Sample Holder,
Multi Pin Holder for 8 Pin Stubs, 3.2mm (1⁄8”)
Expand
View Pricing
933139
SEM Sample Holder,
3-Pin Configuration Holder
Expand
View Pricing
933341
SEM Sample Holder,
SEM Pin Stub Adapter, size 10 mm
Expand
View Pricing
933147
SEM Sample Holder,
5-Pin Configuration Holder
Expand
View Pricing
933104
SEM Sample Holder,
Armray Base Universal Holder
Expand
View Pricing
933163
SEM Sample Holder,
5-Pin Configuration Holder, Pin A
Expand
View Pricing
933295
SEM Sample Holder,
Multiple Holder for 8 Pins Hitachi, M4
Expand
View Pricing