
(S)-(+)-2-Phenylglycine methyl ester hydrochloride

Linear Formula:
C6H5CH(NH2)CO2CH3·HCl
CAS Number:
Molecular Weight:
201.65
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing