Skip to Content
Merck
Sabouraud Dextrose Agar NutriSelect® Plus, Glucose 4 %, powder, bottle of 500 g, suitable for microbiology

Sabouraud Dextrose Agar

Sabouraud Dextrose Agar NutriSelect® Plus, Glucose 4 %, powder, bottle of 500 g, suitable for microbiology

Products

Product Category
Brand
Color
Feature
Physical Form
Agency
Chemical Composition
Capacity (mL)
Diameter (mm)
Compatibility
Quality Segment
Closure Type
Application
Plate Material
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2