
(S)-(−)-4-Methoxy-α-methylbenzylamine

Synonym(s):
(S)-(−)-1-(4-Methoxyphenyl)ethylamine
Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing