Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rubidium chloride
Synonym(s):
Rubidium chloride
Linear Formula:
RbCl
CAS Number:
7791-11-9
Molecular Weight:
120.92
EC Number:
232-240-9
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Isotope
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
R2252
Rubidium chloride,
ReagentPlus
®
, ≥99.0% (metals basis)
Expand
View Pricing
215260
Rubidium chloride,
99.8% trace metals basis
Expand
View Pricing
83979
Rubidium chloride,
BioUltra, for molecular biology, ≥99.0% (AT)
Expand
View Pricing
204250
Rubidium chloride,
99.95% trace metals basis
Expand
View Pricing
1.07615
Rubidium chloride,
for analysis EMSURE
®
Expand
View Pricing
NIST984
Rubidium chloride,
NIST SRM 984
Expand
View Pricing