
Rosiglitazone maleate

Synonym(s):
5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione maleate salt
Empirical Formula (Hill Notation):
C18H19N3O3S · C4H4O4
CAS Number:
Molecular Weight:
473.50
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2