
Rifampicin quinone

Synonym(s):
Dehydrorifampicin
Empirical Formula (Hill Notation):
C43H56N4O12
CAS Number:
Molecular Weight:
820.92
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing