Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reversible 96 well rack
Products
Product Category
racks (6)
Color
Manufacturer
Special Grade
Natural (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
R6151
Reversible 96 well rack,
assorted colors
Expand
View Pricing
R6276
Reversible 96 well rack,
natural
Expand
View Pricing
R6401
Reversible 96 well rack,
blue
Expand
View Pricing
R6776
Reversible 96 well rack,
yellow
Expand
View Pricing
R6651
Reversible 96 well rack,
pink
Expand
View Pricing
R6526
Reversible 96 well rack,
green
Expand
View Pricing