Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reversable Silicone lab mat
Products
Color
Manufacturer
Length (cm)
Color
Package Size
Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
HS120507
Reversable Silicone lab mat,
L × W × thickness 14 in. × 23.5 in. × 2 mm, purple, grey design on one side
Expand
View Pricing
HS120506
Reversable Silicone lab mat,
L × W × thickness 14 in. × 23.5 in. × 2 mm, reversible, yellow, blue design on one side
Expand
View Pricing
HS120747
Reversable Silicone lab mat,
One gridded side, L × W × thickness 14.9 in. × 12 in. × 0.1 in., blue
Expand
View Pricing
HS120748
Reversable Silicone lab mat,
One gridded side, L × W × thickness 14.9 in. × 12 in. × 0.1 in., purple
Expand
View Pricing