Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reagent Bottle, Wide Neck
Products
Product Category
bottles (9)
Capacity (mL)
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
DWK231852404
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231851708
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231882407
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231884408
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231855401
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231853606
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231854405
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231883609
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing
DWK231885404
Reagent Bottle, Wide Neck,
wide-neck
Expand
View Pricing