(R)-1-[(SP)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine
Synonym(s):
(1R)-1-(Dicyclohexylphosphino)-2-[(1R)-1-(dicyclohexylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J003-1
Empirical Formula (Hill Notation):
C36H56FeP2
CAS Number:
Molecular Weight:
606.62
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing