Skip to Content
Merck
Pyruvic acid natural, ≥95%, FG

Pyruvic acid

Pyruvic acid natural, ≥95%, FG
Synonym(s):
α-Ketopropionic acid, 2-Oxopropionic acid, Pyruvic acid
Linear Formula:
CH3COCOOH
CAS Number:
Molecular Weight:
88.06
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing