Pyrimethamine
Synonym(s):
2,4-Diamino-5-(4-chlorophenyl)-6-ethylpyrimidine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine
Empirical Formula (Hill Notation):
C12H13ClN4
CAS Number:
Molecular Weight:
248.71
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique