
Pyridinium chlorochromate

Synonym(s):
PCC
Linear Formula:
C5H6NClCrO3
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing