Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
glass test tubes without rim, medium wall
Products
Product Category
tubes (5)
Manufacturer
Diameter (mm)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z653519
Pyrex
®
glass test tubes without rim, medium wall,
O.D. × L 16 mm × 100 mm, wall thickness 1.2 mm
Expand
View Pricing
Z653489
Pyrex
®
glass test tubes without rim, medium wall,
O.D. × L 10 mm × 75 mm, wall thickness 1.0 mm
Expand
View Pricing
Z653535
Pyrex
®
glass test tubes without rim, medium wall,
O.D. × L 16 mm × 150 mm, wall thickness 1.2 mm
Expand
View Pricing
Z653527
Pyrex
®
glass test tubes without rim, medium wall,
O.D. × L 16 mm × 125 mm, wall thickness 1.2 mm
Expand
View Pricing
Z653543
Pyrex
®
glass test tubes without rim, medium wall,
O.D. × L 18 mm × 150 mm, wall thickness 1.2 mm
Expand
View Pricing