Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Centrifuge tubes
Products
Product Category
tubes (6)
Manufacturer
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SLW3450/02MP
Pyrex
®
Centrifuge tubes,
ASTM Certified, conical base, 100 mL
Expand
View Pricing
SLW3460/02M
Pyrex
®
Centrifuge tubes,
graduated, conical base, 15 mL
Expand
View Pricing
SLW1660/06M
Pyrex
®
Centrifuge tubes,
cylindrical type, round base, 25 mL
Expand
View Pricing
SLW1680/02M
Pyrex
®
Centrifuge tubes,
plain, conical base, 15 mL
Expand
View Pricing
SLW3440/02M
Pyrex
®
Centrifuge tubes,
ASTM Certified, pear-shaped base, 100 mL
Expand
View Pricing
SLW1660/02M
Pyrex
®
Centrifuge tubes,
cylindrical type, round base, 10 mL
Expand
View Pricing