Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pseudoionone
Synonym(s):
ψ-Ionone, 6,10-Dimethyl-3,5,9-undecatrien-2-one
Empirical Formula (Hill Notation):
C
13
H
20
O
CAS Number:
141-10-6
Molecular Weight:
192.30
EC Number:
205-457-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
82536
Pseudoionone,
technical, mixture of isomers, ≥90% (GC)
Expand
View Pricing