Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ProClin
™
950
Synonym(s):
2-Methyl-4-isothiazolin-3-one solution
Products
Product Category
preservatives (6)
Brand
Sigma-Aldrich (6)
Color
pH Value
Physical Form
Chemical Composition
Quality Segment
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
46878-U
ProClin
™
950,
pkg of 50 mL, ProClin™ preservative for use in IVD manufacturing, MIT 9.5 %
Expand
View Pricing
46879-U
ProClin
™
950,
pkg of 400 mL, ProClin
™
preservative for use in IVD manufacturing, MIT 9.5 %
Expand
View Pricing
46885-U
ProClin
™
950,
ProClin
™
preservative for use in IVD manufacturing, pkg of 5 mL, MIT 9.5 %
Expand
View Pricing
46883-U
ProClin
™
950,
pkg of 3.6 L, ProClin
™
preservative for use in IVD manufacturing, MIT 9.5 %
Expand
View Pricing
46884-U
ProClin
™
950,
pkg of 17 L, ProClin
™
preservative for use in IVD manufacturing, MIT 9.5 %
Expand
View Pricing
799130
ProClin
™
950,
pkg of 110 kg, ProClin
™
preservative for use in IVD manufacturing, MIT 9.5 %
Expand
View Pricing