Primidone Related Compound A
Synonym(s):
2-Ethyl-2-phenylmalondiamide, 2-Ethyl-2-phenylpropanediamide, PEMA
Empirical Formula (Hill Notation):
C11H14N2O2
CAS Number:
Molecular Weight:
206.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing