
Pregnenolone

Synonym(s):
3β-Hydroxy-5-pregnen-20-one, 5-Pregnen-3β-ol-20-one
Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing