
Prednisolone sodium phosphate

Synonym(s):
Prednisolone 21-phosphate disodium salt
Empirical Formula (Hill Notation):
C21H27Na2O8P
CAS Number:
Molecular Weight:
484.39
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3