Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Prednisolone 21-hemisuccinate sodium salt
Synonym(s):
Δ
1
-Hydrocortisone 21-hemisuccinate, 1,4-Pregnadiene-11β,17,21-triol-3,20-dione 21-hemisuccinate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-hemisuccinate
Linear Formula:
C
25
H
31
O
8
Na
CAS Number:
1715-33-9
Molecular Weight:
482.50
EC Number:
216-995-1
Products
Product Category
bioactive small molecules (1)
hormones (1)
Brand
Sigma-Aldrich (1)
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Sterility
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P4153
Prednisolone 21-hemisuccinate sodium salt,
≥90%, powder
Expand
View Pricing