
Potassium nonafluoro-1-butanesulfonate

Synonym(s):
Nonafluoro-1-butanesulfonic acid potassium salt, Potassium nonaflate
Linear Formula:
CF3(CF2)3SO3K
CAS Number:
Molecular Weight:
338.19
Products
Product Category
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2