Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinyl chloride)
Synonym(s):
PVC, Poly(vinyl chloride)
Linear Formula:
(CH
2
CHCl)
n
CAS Number:
9002-86-2
Products
Brand
Sigma-Aldrich (5)
Supelco (2)
Manufacturer
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
analytical standard (1)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
81388
Poly(vinyl chloride),
low molecular weight
Expand
View Pricing
81387
Poly(vinyl chloride),
high molecular weight
Expand
View Pricing
346764
Poly(vinyl chloride),
average M
w
~233,000, average M
n
~99,000
Expand
View Pricing
389293
Poly(vinyl chloride),
average M
w
~43,000, average M
n
~22,000
Expand
View Pricing
189588
Poly(vinyl chloride),
average M
w
~62,000, average M
n
~35,000
Expand
View Pricing
1548076
Poly(vinyl chloride),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
81392
Poly(vinyl chloride),
Selectophore
™
, high molecular weight
Expand
View Pricing
182621
Poly(vinyl chloride),
analytical standard, average M
w
85,000 (Typical), average M
n
40,000 (Typical)
Expand
View Pricing
P2600000
Poly(vinyl chloride),
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing