Skip to Content
Merck
Poly(ethylene glycol) methyl ether average Mn 20,000

Poly(ethylene glycol) methyl ether

Poly(ethylene glycol) methyl ether average Mn 20,000
Synonym(s):
Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG
Linear Formula:
CH3(OCH2CH2)nOH
CAS Number:

Products

Product Category
Brand
Melting Point (°C)
pH Value
Physical Form
Functional Group
Polymer Composition
Polymer Type