Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polybutenes
Linear Formula:
[-CH
2
C(CH
3
)
2
-]
x
[-CH
2
CH(C
2
H
5
)-]
y
CAS Number:
9003-29-6
Products
Brand
Sigma-Aldrich (2)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Polymer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
388696
Polybutenes,
average M
n
~920 by VPO, isobutylene >90 %
Expand
View Pricing
388726
Polybutenes,
average M
n
~2,300 by VPO, isobutylene >90 %
Expand
View Pricing