
Podophyllotoxin for system suitability

Synonym(s):
Podophyllotoxin
Empirical Formula (Hill Notation):
C22H22O8
CAS Number:
Molecular Weight:
414.41
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing