Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum nanospheres
Empirical Formula (Hill Notation):
Pt
CAS Number:
7440-06-4
Molecular Weight:
195.08
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Quality Segment
Functional Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
916056
Platinum nanospheres,
citrate functionalized, 5 nm diameter, 0.05 mg/mL (2 mM aqueous sodium citrate buffer)
Expand
View Pricing
915041
Platinum nanospheres,
citrate functionalized, 50 nm diameter, 0.05 mg/mL in 2 mM aqueous sodium citrate buffer
Expand
View Pricing
916307
Platinum nanospheres,
citrate functionalized, 30 nm diameter, 0.05 mg/mL (2 mM aqueous sodium citrate buffer)
Expand
View Pricing