Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips for Eppendorf
®
and Socorex pipettors
Products
Product Category
pipette tips (2)
Color
Manufacturer
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
AXYTE1004B
Pipette tips for Eppendorf
®
and Socorex pipettors,
size 1000 μL, blue, pkg of 10x500tips/cs
Expand
View Pricing
AXYTE1004BR
Pipette tips for Eppendorf
®
and Socorex pipettors,
racked, size 1000 μL, blue, pkg of 3x1152tips/cs
Expand
View Pricing