Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Piperidine
Synonym(s):
Piperidine, Hexahydropyridine
Empirical Formula (Hill Notation):
C
5
H
11
N
CAS Number:
110-89-4
Molecular Weight:
85.15
EC Number:
203-813-0
Products
Product Category
solvents (2)
aroma chemicals (1)
organic bases (1)
Brand
Sigma-Aldrich (5)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Biotechnology (1)
Food Grade (1)
Halal (1)
Kosher (1)
analytical standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
8.22299
Piperidine,
for synthesis
Expand
View Pricing
411027
Piperidine,
≥99.5%, purified by redistillation
Expand
View Pricing
571261
Piperidine,
biotech. grade, ≥99.5%
Expand
View Pricing
104094
Piperidine,
ReagentPlus
®
, 99%
Expand
View Pricing
1.09724
Piperidine,
for analysis EMSURE
®
Expand
View Pricing
W290820
Piperidine,
≥99%, FG
Expand
View Pricing
76046
Piperidine,
analytical standard
Expand
View Pricing