Skip to Content
Merck
Piperazine BioUltra, anhydrous, ≥99.0% (T)

Piperazine

Piperazine BioUltra, anhydrous, ≥99.0% (T)
Synonym(s):
1,4-Diazacyclohexane, Diethylenediamine
Empirical Formula (Hill Notation):
C4H10N2
CAS Number:
Molecular Weight:
86.14
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing